missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanobenzyl alcohol, 97%

Manufacturer:  Alfa Aesar™ H54283MD

 View more versions of this product

Catalog No. AAH54283MD



Description

Description

Applications
4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-Cyanobenzyl alcohol
250mg
C8H7NO
UN3439
Insoluble in water.
C1=CC(=CC=C1CO)C#N
133.15
133.15
32°C to 36°C
874-89-5
MFCD00870633
4 hydroxymethyl-benzonitrile, 4-cyanobenzenemethanol, 4-cyanobenzyl alcohol, 4-cyanophenyl methanol, 4-hydroxymethyl benzonitrile, 4-hydroxymethyl-benzonitrile, benzonitrile, 4-hydroxymethyl, ccris 5116, p-cyanobenzyl alcohol
XAASLEJRGFPHEV-UHFFFAOYSA-N
4-(hydroxymethyl)benzonitrile
160549
97%

Chemical Identifiers

874-89-5
133.15
XAASLEJRGFPHEV-UHFFFAOYSA-N
160549
C1=CC(=CC=C1CO)C#N
C8H7NO
MFCD00870633
4 hydroxymethyl-benzonitrile, 4-cyanobenzenemethanol, 4-cyanobenzyl alcohol, 4-cyanophenyl methanol, 4-hydroxymethyl benzonitrile, 4-hydroxymethyl-benzonitrile, benzonitrile, 4-hydroxymethyl, ccris 5116, p-cyanobenzyl alcohol
4-(hydroxymethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents