Learn More
4-Cyanobenzoic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 110730250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Cyanobenzoic acid | |
619-65-8 | |
100.0 | |
Beige-Yellow to White | |
98.5% min. (HPLC) | |
C8H5NO2 | |
MFCD00002528 | |
09, 845 | |
Solubility in water: 1.2g/L (20°C). Other solubilities: soluble in methanol | |
C1=CC(=CC=C1C#N)C(=O)O | |
147.13 | |
147.13 | |
Crystalline Powder |
99% | |
98.5 | |
217.0°C to 222.0°C | |
Authentic | |
Glass bottle | |
NCC6H4CO2H | |
25 g | |
p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acid | |
ADCUEPOHPCPMCE-UHFFFAOYSA-N | |
4-cyanobenzoic acid | |
12087 | |
99% |
Chemical Identifiers
619-65-8 | |
147.13 | |
ADCUEPOHPCPMCE-UHFFFAOYSA-N | |
12087 | |
C1=CC(=CC=C1C#N)C(=O)O |
C8H5NO2 | |
MFCD00002528 | |
p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acid | |
4-cyanobenzoic acid |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
GHS Signal Word: Warning
EINECSNumber : 210-606-9
TSCA : TSCA
RUO – Research Use Only