missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanobenzaldehyde, 98+%

Manufacturer:  Alfa Aesar™ A1491422

 View more versions of this product

Catalog No. AAA1491422



Description

Description

Applications
4-Cyanobenzaldehyde, is used as an intermediate for organic synthesis.It is also used as an intermediate in the pharmaceutical and research department.

Solubility
It is soluble in water, ethanol, ether and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong acids, Strong bases, Strong reducing agents.
Specifications

Specifications

4-Cyanobenzaldehyde
133°C (12mmHg)
100g
C8H5NO
606473
WZWIQYMTQZCSKI-UHFFFAOYSA-N
4-formylbenzonitrile
66042
≥98%
Characteristic
99°C to 103°C
105-07-7
MFCD00003376
4-cyanobenzaldehyde, benzonitrile, 4-formyl, p-cyanobenzaldehyde, p-formylbenzonitrile, benzaldehyde, p-cyano, benzonitrile, p-formyl, usaf kf-1, terephthalaldehydonitrile, p-cyanobenzenecarboxaldehyde, 4-formyl-benzonitrile
C1=CC(=CC=C1C=O)C#N
131.134
131.13

Chemical Identifiers

105-07-7
131.134
WZWIQYMTQZCSKI-UHFFFAOYSA-N
66042
C1=CC(=CC=C1C=O)C#N
C8H5NO
MFCD00003376
4-cyanobenzaldehyde, benzonitrile, 4-formyl, p-cyanobenzaldehyde, p-formylbenzonitrile, benzaldehyde, p-cyano, benzonitrile, p-formyl, usaf kf-1, terephthalaldehydonitrile, p-cyanobenzenecarboxaldehyde, 4-formyl-benzonitrile
4-formylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : CU5250000

TSCA : TSCA

SDS Documents