Learn More
4-Cyanobenzaldehyde, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1491414
Description
4-Cyanobenzaldehyde, is used as an intermediate for organic synthesis.It is also used as an intermediate in the pharmaceutical and research department.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cyanobenzaldehyde, is used as an intermediate for organic synthesis.It is also used as an intermediate in the pharmaceutical and research department.
Solubility
It is soluble in water, ethanol, ether and chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong acids, Strong bases, Strong reducing agents.
Specifications
4-Cyanobenzaldehyde | |
99°C to 103°C | |
Characteristic | |
MFCD00003376 | |
606473 | |
4-cyanobenzaldehyde, benzonitrile, 4-formyl, p-cyanobenzaldehyde, p-formylbenzonitrile, benzaldehyde, p-cyano, benzonitrile, p-formyl, usaf kf-1, terephthalaldehydonitrile, p-cyanobenzenecarboxaldehyde, 4-formyl-benzonitrile | |
WZWIQYMTQZCSKI-UHFFFAOYSA-N | |
4-formylbenzonitrile | |
66042 | |
≥98% |
105-07-7 | |
133°C (12 mmHg) | |
C8H5NO | |
25 g | |
Air Sensitive | |
It is soluble in water,ethanol,ether and chloroform. | |
O=CC1=CC=C(C=C1)C#N | |
131.13 | |
131.13 |
Chemical Identifiers
105-07-7 | |
131.13 | |
WZWIQYMTQZCSKI-UHFFFAOYSA-N | |
66042 | |
O=CC1=CC=C(C=C1)C#N |
C8H5NO | |
MFCD00003376 | |
4-cyanobenzaldehyde, benzonitrile, 4-formyl, p-cyanobenzaldehyde, p-formylbenzonitrile, benzaldehyde, p-cyano, benzonitrile, p-formyl, usaf kf-1, terephthalaldehydonitrile, p-cyanobenzenecarboxaldehyde, 4-formyl-benzonitrile | |
4-formylbenzonitrile |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 203-267-3
RTECSNumber : CU5250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only