missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyano-D-phenylalanine, 98%

Manufacturer:  Alfa Aesar™ H6368706

 View more versions of this product

Catalog No. AAH6368706



Description

Description

Applications
4-Cyano-D-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

4-Cyano-D-phenylalanine
C10H10N2O2
UN3439
KWIPUXXIFQQMKN-SECBINFHSA-N
(2R)-2-amino-3-(4-cyanophenyl)propanoic acid
1501864
Solid
White
263396-44-7
MFCD00270364
r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cyanophenylalanine, h-d-phe 4-cn-oh, 4-cyano-d-phenylalanine, 2r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cn-phe-oh, d-4-cyanophe, h-4-cyano-phe-oh, ambotzhaa7570, pubchem13160
C1=CC(=CC=C1CC(C(=O)O)N)C#N
190.202
190.2
98%
5g

Chemical Identifiers

263396-44-7
190.202
KWIPUXXIFQQMKN-SECBINFHSA-N
1501864
C1=CC(=CC=C1CC(C(=O)O)N)C#N
C10H10N2O2
MFCD00270364
r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cyanophenylalanine, h-d-phe 4-cn-oh, 4-cyano-d-phenylalanine, 2r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cn-phe-oh, d-4-cyanophe, h-4-cyano-phe-oh, ambotzhaa7570, pubchem13160
(2R)-2-amino-3-(4-cyanophenyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

storageNote1 : -30° to -10°C

SDS Documents