missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyano-4-phenylpiperidine Hydrochloride 99%, ACROS Organics™

$164.89 - $164.89

Chemical Identifiers

CAS 51304-58-6
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C12H14N2·HCl
Formula Weight 222.72
MDL Number MFCD00012775
Synonym 4-Phenyl-4-piperidinecarbonitrile hydrochloride
Chemical Name or Material 4-Cyano-4-phenylpiperidine hydrochloride, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC110840500 SDS Thermo Fisher Scientific
110840500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC110840100 SDS Thermo Fisher Scientific
110840100
Glass bottle 10g Each for $164.89
Specifications Description & Specifications

Chemical Identifiers

51304-58-6
100.0
C12H14N2·HCl
MFCD00012775
4-Cyano-4-phenylpiperidine hydrochloride, 99%
98.5
99%
222.72
4-Phenyl-4-piperidinecarbonitrile hydrochloride
≥98.5%

Specifications

222.72
50g
210° to 214°C
Authentic
Glass bottle
99%
51304-58-6
4-Cyano-4-phenylpiperidine hydrochloride, 99%
Crystalline Powder
White to off-white
100.0
4-Phenyl-4-piperidinecarbonitrile hydrochloride
MFCD00012775
98.5
C12H14N2·HCl
≥98.5%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 257-124-5