Please login to your online account to display your discounted pricing

4-Cyano-4-phenylpiperidine Hydrochloride 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  110840100

 View more versions of this product

Catalog No. AC110840100

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 222.72
Physical Form Crystalline Powder
Assay ≥98.5%
Quantity 10g
Color White to off-white
EINECS Number 257-124-5
Melting Point 210° to 214°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Name Note 99%
CAS 51304-58-6
Assay Percent Range 99%
Molecular Formula C12H14N2·HCl
Chemical Name or Material 4-Cyano-4-phenylpiperidine hydrochloride
MDL Number MFCD00012775
CAS Min % 98.5
Infrared Spectrum Authentic
Synonym 4-Phenyl-4-piperidinecarbonitrile hydrochloride
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 4-Cyano-4-phenylpiperidine hydrochloride
Name Note 99%
CAS 51304-58-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H14N2·HCl
Formula Weight 222.72
MDL Number MFCD00012775
Synonym 4-Phenyl-4-piperidinecarbonitrile hydrochloride
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents