Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyano-4-phenylcyclohexanone, 98%

$175.47 - $175.47

Chemical Identifiers

Chemical Name or Material 4-Cyano-4-phenylcyclohexanone
CAS 25115-74-6
Molecular Formula C13H13NO
Formula Weight 199.24
MDL Number MFCD00044816
Beilstein 2645593
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0528106

 
alfa aesar™
L0528106
5g Each for $175.47
Description & Specifications

Specifications

Formula Weight 199.24
Chemical Name or Material 4-Cyano-4-phenylcyclohexanone
Assay 98%
Molecular Formula C13H13NO
MDL Number MFCD00044816
Beilstein 2645593
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
CAS 25115-74-6
Melting Point 112° to 118°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.