missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyano-4'-n-octyloxybiphenyl, 97%

Manufacturer:  Alfa Aesar™ H5519103

 View more versions of this product

Catalog No. AAH5519103



Specifications

Specifications

4-Cyano-4'-n-octyloxybiphenyl
51°C to 77°C
52364-73-5
MFCD00075145
GPGGNNIMKOVSAG-UHFFFAOYSA-N
4-(4-octoxyphenyl)benzonitrile
104173
97%
113°C (235°F)
1g
C21H25NO
1,1'-biphenyl-4-carbonitrile, 4'-octyloxy
CCCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
307.437
307.44

Chemical Identifiers

52364-73-5
307.437
GPGGNNIMKOVSAG-UHFFFAOYSA-N
104173
CCCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
C21H25NO
MFCD00075145
1,1'-biphenyl-4-carbonitrile, 4'-octyloxy
4-(4-octoxyphenyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DV1764500

TSCA : TSCA

Documents