missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyano-4'-n-hexyloxybiphenyl, 97%

Manufacturer:  Alfa Aesar™ H5587603

 View more versions of this product

Catalog No. AAH5587603



Description

Description

Applications
4-Cyano-4′-n-hexyloxybiphenyl, is used as a intermediate of liquid crystals. It is mainly used as the pharmaceutical intermediate, liquid crystal intermediate, liquid crystal monomers.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

4-Cyano-4'-n-hexyloxybiphenyl
1g
C19H21NO
1,1'-biphenyl-4-carbonitrile, 4'-hexyloxy
YUYXUPYNSOFWFV-UHFFFAOYSA-N
4-(4-hexoxyphenyl)benzonitrile
162465
97%
56°C to 58°C
41424-11-7
MFCD00134145
Slightly soluble in water.
CCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
279.383
279.38

Chemical Identifiers

41424-11-7
279.383
YUYXUPYNSOFWFV-UHFFFAOYSA-N
162465
CCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
C19H21NO
MFCD00134145
1,1'-biphenyl-4-carbonitrile, 4'-hexyloxy
4-(4-hexoxyphenyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 255-358-2

RTECSNumber : DV1764200

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents