Learn More
4-Cyano-4'-n-hexylbiphenyl, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H55825MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-Cyano-4′-n-hexylbiphenyl, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a liquid crystal monomer.
Solubility
Sparingly Soluble in water (3.2E-5 g/L) (25°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
4-Cyano-4'-n-hexylbiphenyl | |
113°C (235°F) | |
MFCD00075144 | |
4-hexyl-4'-cyanobiphenyl, 4-cyano-4'-hexylbiphenyl, 4'-hexyl-4-biphenylcarbonitrile, 4-n-hexyl-4'-cyanobiphenyl, 4'-hexyl-1,1'-biphenyl-4-carbonitrile, 4'-hexylbiphenyl-4-carbonitrile, ccris 6044, 1,1'-biphenyl-4-carbonitrile, 4'-hexyl, 4-4-hexylphenyl benzonitrile | |
VADSDVGLFDVIMG-UHFFFAOYSA-N | |
4-(4-hexylphenyl)benzonitrile | |
38764 | |
97% |
41122-70-7 | |
C19H21N | |
250 mg | |
Sparingly Soluble in water (3.2E-5g/L) (25°C). | |
CCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N | |
263.384 | |
263.38 |
Chemical Identifiers
41122-70-7 | |
263.384 | |
VADSDVGLFDVIMG-UHFFFAOYSA-N | |
38764 | |
CCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N |
C19H21N | |
MFCD00075144 | |
4-hexyl-4'-cyanobiphenyl, 4-cyano-4'-hexylbiphenyl, 4'-hexyl-4-biphenylcarbonitrile, 4-n-hexyl-4'-cyanobiphenyl, 4'-hexyl-1,1'-biphenyl-4-carbonitrile, 4'-hexylbiphenyl-4-carbonitrile, ccris 6044, 1,1'-biphenyl-4-carbonitrile, 4'-hexyl, 4-4-hexylphenyl benzonitrile | |
4-(4-hexylphenyl)benzonitrile |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 255-228-5
TSCA : Yes
Recommended Storage : Ambient temperatures