Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyano-4'-n-heptyloxybiphenyl, 99%

$334.50 - $334.50

Chemical Identifiers

Chemical Name or Material 4-Cyano-4'-n-heptyloxybiphenyl
CAS 52364-72-4
Molecular Formula C20H23NO
Formula Weight 293.41
MDL Number MFCD00134146
Synonym 4'-n-Heptyloxy-4-biphenylcarbonitrile; 4-(4'-n-Heptyloxyphenyl)benzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2204406

  SDS
alfa aesar™
B2204406
5g Each for $334.50
Description & Specifications

Specifications

Formula Weight 293.41
Assay 99%
CAS 52364-72-4
MDL Number MFCD00134146
Synonym 4'-n-Heptyloxy-4-biphenylcarbonitrile; 4-(4'-n-Heptyloxyphenyl)benzonitrile
Melting Point 53° to 55°C
Quantity 5g
RTECS Number DV1764100
Chemical Name or Material 4-Cyano-4'-n-heptyloxybiphenyl
Molecular Formula C20H23NO
TSCA TSCA