missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Cyano-4-biphenylyl trans-4-ethylcyclohexanecarboxylate, 97%

Manufacturer:  Alfa Aesar™ H5505403

 View more versions of this product

Catalog No. AAH5505403



Specifications

Specifications

4'-Cyano-4-biphenylyl trans-4-ethylcyclohexanecarboxylate
1g
C22H23NO2
4'-cyano-1,1'-biphenyl-4-yl 1s,4r-4-ethylcyclohexane-1-carboxylate
CCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)C3=CC=C(C=C3)C#N
333.431
333.43
83°C to 86°C
67284-56-4
MFCD09751067
OYZITXKNUGHHKV-UHFFFAOYSA-N
[4-(4-cyanophenyl)phenyl] 4-ethylcyclohexane-1-carboxylate
171826
97%

Chemical Identifiers

67284-56-4
333.431
OYZITXKNUGHHKV-UHFFFAOYSA-N
171826
CCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)C3=CC=C(C=C3)C#N
C22H23NO2
MFCD09751067
4'-cyano-1,1'-biphenyl-4-yl 1s,4r-4-ethylcyclohexane-1-carboxylate
[4-(4-cyanophenyl)phenyl] 4-ethylcyclohexane-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 266-631-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents