missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyano-3-fluorophenyl 4-Butylbenzoate, TCI America™

Manufacturer:  TCI America C33415G

 View more versions of this product

Catalog No. C33415G



Specifications

Specifications

4-Cyano-3-fluorophenyl 4-Butylbenzoate
8°C
86776-52-5
MFCD11053362
IBUUMDHMTARCSQ-UHFFFAOYSA-N
(4-cyano-3-fluorophenyl) 4-butylbenzoate
13742342
Clear Liquid-Cloudy Liquid at 20°C
Yellow
5g
C18H16FNO2
4-butylbenzoic acid 4-cyano-3-fluorophenyl ester, 4-butyl-benzoic acid 4-cyano-3-fluoro-phenyl ester, 4-cyano-3-fluorophenyl=4-butylbenzoate, 4-cyano-3-fluorophenyl 4-n-butylbenzoate, 4-cyano-3-fluorophenyl 4-n-butylbenzoate, benzoic acid, 4-butyl-, 4-cyano-3-fluorophenyl ester, benzoic acid, 4-butyl-,4-cyano-3-fluorophenyl ester
CCCCC1=CC=C(C=C1)C(=O)OC2=CC(=C(C=C2)C#N)F
297.329
297.33
≥98.0% (GC)

Chemical Identifiers

86776-52-5
297.329
IBUUMDHMTARCSQ-UHFFFAOYSA-N
13742342
CCCCC1=CC=C(C=C1)C(=O)OC2=CC(=C(C=C2)C#N)F
C18H16FNO2
MFCD11053362
4-butylbenzoic acid 4-cyano-3-fluorophenyl ester, 4-butyl-benzoic acid 4-cyano-3-fluoro-phenyl ester, 4-cyano-3-fluorophenyl=4-butylbenzoate, 4-cyano-3-fluorophenyl 4-n-butylbenzoate, 4-cyano-3-fluorophenyl 4-n-butylbenzoate, benzoic acid, 4-butyl-, 4-cyano-3-fluorophenyl ester, benzoic acid, 4-butyl-,4-cyano-3-fluorophenyl ester
(4-cyano-3-fluorophenyl) 4-butylbenzoate
Safety and Handling

Safety and Handling

TSCA : No

Documents