missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyano-3-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H62377MD

 View more versions of this product

Catalog No. AAH62377MD



Specifications

Specifications

4-Cyano-3-fluorobenzeneboronic acid pinacol ester
870238-67-8
MFCD16996321
2-4-cyano-3-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C#N)F
247.076
247.07
96%
250mg
C13H15BFNO2
14562802
DMJGKVYFBOCENL-UHFFFAOYSA-N
2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
51350951
Solid

Chemical Identifiers

870238-67-8
247.076
DMJGKVYFBOCENL-UHFFFAOYSA-N
51350951
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C#N)F
C13H15BFNO2
MFCD16996321
2-4-cyano-3-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents