missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyano-3,5-difluoropyridine, 98%

Chemical Identifiers

CAS 1214377-09-9
Formula Weight 140.09
MDL Number MFCD13176565
UN Number UN3439
Synonym 3,5-Difluoropyridine-4-carbonitrile
Chemical Name or Material 4-Cyano-3,5-difluoropyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6439103 SDS Alfa Aesar™
H6439103
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH64391MD SDS Alfa Aesar™
H64391MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1214377-09-9
MFCD13176565
3,5-Difluoropyridine-4-carbonitrile
140.09
UN3439
4-Cyano-3,5-difluoropyridine

Specifications

1g
Solid
MFCD13176565
3,5-Difluoropyridine-4-carbonitrile
4-Cyano-3,5-difluoropyridine
1214377-09-9
140.09
UN3439
Off-white
98%

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.