missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyano-2-iodopyridine, 95%

$227.81 - $760.41

Chemical Identifiers

CAS 114821-24-8
Molecular Formula C6H3IN2
Formula Weight 230.01
MDL Number MFCD08689485
Synonym 2-Iodoisonocotinonitrile; 2-Iodopyridine-4-carbonitrile
Chemical Name or Material 4-Cyano-2-iodopyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3106503 SDS Alfa Aesar™
H3106503
1g Each for $227.81
AAH3106506 SDS Alfa Aesar™
H3106506
5g Each for $760.41
Specifications Description & Specifications

Chemical Identifiers

114821-24-8
230.01
2-Iodoisonocotinonitrile; 2-Iodopyridine-4-carbonitrile
C6H3IN2
MFCD08689485
4-Cyano-2-iodopyridine

Specifications

77° to 80°C
114821-24-8
230.01
2-Iodoisonocotinonitrile; 2-Iodopyridine-4-carbonitrile
95%
1g
C6H3IN2
MFCD08689485
4-Cyano-2-iodopyridine

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning