Learn More
4-Cyano-2-fluoropyridine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2593806
Description
4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with hydrogen fluoride, nitrogen oxides, strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Specifications
4-Cyano-2-fluoropyridine | |
31°C to 33°C | |
MFCD09027299 | |
2-fluoroisonicotinonitrile, 4-cyano-2-fluoropyridine, 2-fluoro-4-cyanopyridine, 4-pyridinecarbonitrile, 2-fluoro, 2-fluoro-4-pyridinecarbonitrile, 2-fluoro-isonicotinonitrile, 4-pyridinecarbonitrile,2-fluoro, acmc-1bnd8, 4-cyano-2-fluoro pyridine, ksc581g3h | |
DYQMGPSBUBTNJZ-UHFFFAOYSA-N | |
2-fluoropyridine-4-carbonitrile | |
12002587 | |
97% |
3939-14-8 | |
C6H3FN2 | |
5 g | |
Insoluble in water. | |
C1=CN=C(C=C1C#N)F | |
122.102 | |
122.1 |
Chemical Identifiers
3939-14-8 | |
122.102 | |
DYQMGPSBUBTNJZ-UHFFFAOYSA-N | |
2-fluoropyridine-4-carbonitrile |
C6H3FN2 | |
MFCD09027299 | |
12002587 | |
C1=CN=C(C=C1C#N)F |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only