Learn More
4-Cyano-2-fluorobenzoic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 164149-28-4 |
---|---|
Molecular Formula | C8H4FNO2 |
Molecular Weight (g/mol) | 165.12 |
MDL Number | MFCD03094454 |
InChI Key | KEJMSTJTAWACNI-UHFFFAOYSA-N |
Synonym | 2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile |
PubChem CID | 590659 |
IUPAC Name | 4-cyano-2-fluorobenzoic acid |
SMILES | OC(=O)C1=C(F)C=C(C=C1)C#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC441830010
|
Thermo Scientific Chemicals
441830010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
164149-28-4 | |
165.12 | |
KEJMSTJTAWACNI-UHFFFAOYSA-N | |
590659 | |
OC(=O)C1=C(F)C=C(C=C1)C#N |
C8H4FNO2 | |
MFCD03094454 | |
2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile | |
4-cyano-2-fluorobenzoic acid |
Specifications
164149-28-4 | |
100.0 | |
96% min. (HPLC) | |
C8H4FNO2 | |
1g | |
KEJMSTJTAWACNI-UHFFFAOYSA-N | |
4-cyano-2-fluorobenzoic acid | |
590659 | |
97% |
96.0 | |
Authentic | |
Glass bottle | |
MFCD03094454 | |
2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile | |
OC(=O)C1=C(F)C=C(C=C1)C#N | |
165.12 | |
165.12 | |
4-Cyano-2-fluorobenzoic acid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Contaminated work clothing should not be allowed out of the workplace.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only