missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyano-2-fluorobenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H61134MD

 View more versions of this product

Catalog No. AAH61134MD



Description

Description

Applications
4-Cyano-2-fluorobenzeneboronic acid is used an active pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-Cyano-2-fluorobenzeneboronic acid
1150114-77-4
MFCD07644472
HFAUMAIVCYTVQR-UHFFFAOYSA-N
(4-cyano-2-fluorophenyl)boronic acid
44755209
Solid
250mg
C7H5BFNO2
4-borono-3-fluorobenzonitrile, 4-cyano-2-fluoro-phenylboronic acid, 4-cyano-2-fluorobenzeneboronic acid, 4-cyano-2-fluorophenyl boronic acid, acmc-2099nb, boronic acid, b-4-cyano-2-fluorophenyl
B(C1=C(C=C(C=C1)C#N)F)(O)O
164.93
164.93
98%

Chemical Identifiers

1150114-77-4
164.93
HFAUMAIVCYTVQR-UHFFFAOYSA-N
44755209
B(C1=C(C=C(C=C1)C#N)F)(O)O
C7H5BFNO2
MFCD07644472
4-borono-3-fluorobenzonitrile, 4-cyano-2-fluoro-phenylboronic acid, 4-cyano-2-fluorobenzeneboronic acid, 4-cyano-2-fluorophenyl boronic acid, acmc-2099nb, boronic acid, b-4-cyano-2-fluorophenyl
(4-cyano-2-fluorophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents