Please login to your online account to display your discounted pricing

4-Cumylphenol 99%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  110645000

 View more versions of this product

Catalog No. AC110645000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Needles
Assay ≥98.5%
Formula Weight 212.29
Quantity 500g
Color White
EINECS Number 209-968-0
TSCA TSCA
Melting Point 72° to 75°C
RTECS Number SL1942450
CAS Max % 100.0
Chemical Name or Material 4-Cumylphenol
Name Note 99%
Packaging Plastic bottle
Assay Percent Range 99%
Molecular Formula C15H16O
Linear Formula C6H5C(CH3)2C6H4OH
MDL Number MFCD00002365
CAS 599-64-4
Synonym 4-(2-Phenylisopropyl)phenol
CAS Min % 98.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Cumylphenol
Name Note 99%
CAS 599-64-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C15H16O
Linear Formula C6H5C(CH3)2C6H4OH
Formula Weight 212.29
MDL Number MFCD00002365
Synonym 4-(2-Phenylisopropyl)phenol