missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cumylphenol 99%, ACROS Organics™

Chemical Identifiers

CAS 599-64-4
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C15H16O
Linear Formula C6H5C(CH3)2C6H4OH
Formula Weight 212.29
MDL Number MFCD00002365
Synonym 4-(2-Phenylisopropyl)phenol
Chemical Name or Material 4-Cumylphenol, 99%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC110645000 SDS Thermo Fisher Scientific
110645000
Plastic bottle 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

599-64-4
100.0
C15H16O
212.29
4-(2-Phenylisopropyl)phenol
≥98.5%
98.5
99%
C6H5C(CH3)2C6H4OH
MFCD00002365
4-Cumylphenol, 99%

Specifications

212.29
500g
72° to 75°C
Plastic bottle
C15H16O
599-64-4
C6H5C(CH3)2C6H4OH
Authentic
≥98.5%
Fine Crystalline Needles
White
100.0
4-(2-Phenylisopropyl)phenol
99%
98.5
MFCD00002365
4-Cumylphenol, 99%

Safety and Handling

EINECSNumber : 209-968-0

RTECSNumber : SL1942450

TSCA : TSCA