Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-cis-Fluoro-L-prolinamide hydrochloride, 98%

$135.77 - $338.64

Chemical Identifiers

Chemical Name or Material 4-cis-Fluoro-L-prolinamide hydrochloride
CAS 426844-23-7
Molecular Formula C5H9FN2O·HCl
Formula Weight 168.59
MDL Number MFCD07368567
Synonym (2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27235MD

  SDS
alfa aesar™
H27235MD
250mg Each for $135.77

AAH2723503

  SDS
alfa aesar™
H2723503
1g Each for $338.64
Description & Specifications

Specifications

Formula Weight 168.59
CAS 426844-23-7
Quantity 250mg
Optical Rotation −45 (c=1 in Water)
Chemical Name or Material 4-cis-Fluoro-L-prolinamide hydrochloride
Assay 98%
Molecular Formula C5H9FN2O·HCl
MDL Number MFCD07368567
Synonym (2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 231° to 236°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.