missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chromanone, 96%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 153460500

 View more versions of this product

Catalog No. AC153460500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Chromanone
Authentic
1.575
White to Yellow
50g
491-37-2
100.0
C9H8O2
17, II, 335
MSTDXOZUKAQDRL-UHFFFAOYSA-N
2,3-dihydrochromen-4-one
68110
Crystalline Powder, Crystals or Low Melting Mass
110°C
Glass bottle
127°C to 128°C (13.0mmHg)
34°C to 40°C
96%
95.0
95% min. (GC)
MFCD00006840
2,3-dihydro-1-benzopyran-4-one
C1COC2=CC=CC=C2C1=O
148.16
148.16
96%

Chemical Identifiers

491-37-2
148.16
MSTDXOZUKAQDRL-UHFFFAOYSA-N
68110
C1COC2=CC=CC=C2C1=O
C9H8O2
MFCD00006840
2,3-dihydro-1-benzopyran-4-one
2,3-dihydrochromen-4-one
Safety and Handling

Safety and Handling

EINECSNumber : 207-736-3

RTECSNumber : DJ2981225

TSCA : TSCA

SDS Documents