missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chromanone, 96%, Thermo Scientific™
Chemical Identifiers
CAS | 491-37-2 |
---|---|
Molecular Formula | C9H8O2 |
Molecular Weight (g/mol) | 148.16 |
MDL Number | MFCD00006840 |
InChI Key | MSTDXOZUKAQDRL-UHFFFAOYSA-N |
Synonym | 4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone |
PubChem CID | 68110 |
IUPAC Name | 2,3-dihydrochromen-4-one |
SMILES | C1COC2=CC=CC=C2C1=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC153460100
|
Thermo Scientific Chemicals
153460100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC153460500
|
Thermo Scientific Chemicals
153460500 |
50g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
491-37-2 | |
148.16 | |
MSTDXOZUKAQDRL-UHFFFAOYSA-N | |
68110 | |
C1COC2=CC=CC=C2C1=O |
C9H8O2 | |
MFCD00006840 | |
4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone | |
2,3-dihydrochromen-4-one |
Specifications
491-37-2 | |
100.0 | |
110°C | |
95% min. (GC) | |
C9H8O2 | |
MFCD00006840 | |
17, II, 335 | |
MSTDXOZUKAQDRL-UHFFFAOYSA-N | |
2,3-dihydrochromen-4-one | |
68110 | |
96% |
95.0 | |
127°C to 128°C (13.0mmHg) | |
Authentic | |
Glass bottle | |
1.575 | |
10g | |
4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone | |
C1COC2=CC=CC=C2C1=O | |
148.16 | |
148.16 | |
4-Chromanone, 96% |
Safety and Handling
EINECSNumber : 207-736-3
RTECSNumber : DJ2981225
TSCA : TSCA
RUO â Research Use Only