Please login to your online account to display your discounted pricing

4-Cholesten-3-one, ≥98%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  182420050

 View more versions of this product

Catalog No. AC182420050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Crystalline Powder
Formula Weight 384.64
Quantity 5g
Color White to slightly yellow
EINECS Number 210-005-1
TSCA TSCA
RTECS Number FZ7700000
Melting Point 79° to 82°C
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 4-Cholesten-3-one
Specific Rotation +90° to +95° (20°C, 589nm) (c=2, CHCl3)
Name Note 99.5+%
Assay Percent Range 98+%
Molecular Formula C27H44O
CAS 601-57-0
Synonym 3-Oxo-4-cholestene
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Cholesten-3-one
Name Note 99.5+%
CAS 601-57-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C27H44O
Formula Weight 384.64
Synonym 3-Oxo-4-cholestene