missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cholesten-3-one, ≥98%, Acros Organics

Chemical Identifiers

CAS 601-57-0
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C27H44O
Formula Weight 384.64
Synonym 3-Oxo-4-cholestene
Chemical Name or Material 4-Cholesten-3-one, 99.5+%
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC182420050 SDS Thermo Fisher Scientific
182420050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

601-57-0
100.0
C27H44O
3-Oxo-4-cholestene
≥98.0%
98.0
98+%
384.64
4-Cholesten-3-one, 99.5+%

Specifications

384.64
5g
79° to 82°C
100.0
+90° to +95° (20°C, 589nm) (c=2, CHCl3)
98+%
Glass bottle
4-Cholesten-3-one, 99.5+%
Crystalline Powder
White to slightly yellow
98.0
3-Oxo-4-cholestene
C27H44O
601-57-0
Authentic
≥98.0%

Safety and Handling

EINECSNumber : 210-005-1

RTECSNumber : FZ7700000

TSCA : TSCA