missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cholesten-3-one, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1174306

 View more versions of this product

Catalog No. AAA1174306



Description

Description

4-Cholesten-3-one is used as a chiral building block for proteomics research. It is an intestinal metabolite of cholesterol and exhibits an anti-obesity effect on animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cholesten-3-one is used as a chiral building block for proteomics research. It is an intestinal metabolite of cholesterol and exhibits an anti-obesity effect on animals.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Cholesten-3-one
79°C to 81°C
MFCD00003663
4707774
Slightly soluble in water.
+93° (c=1 in chloroform)
(8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
91477
384.65
601-57-0
C27H44O
5 g
4-cholesten-3-one, cholest-4-en-3-one, cholestenone, 3-oxocholest-4-ene, delta sup 4-cholestenone, +-4-cholesten-3-one, unii-7t94nhd99c, 3-oxo-4-cholestene, 3-keto-4-cholestene, cholestenone delta 4
NYOXRYYXRWJDKP-GYKMGIIDSA-N
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C
384.648
CHEBI:16175
≥98%

Chemical Identifiers

601-57-0
384.648
NYOXRYYXRWJDKP-GYKMGIIDSA-N
91477
(8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
C27H44O
MFCD00003663
4-cholesten-3-one, cholest-4-en-3-one, cholestenone, 3-oxocholest-4-ene, delta sup 4-cholestenone, +-4-cholesten-3-one, unii-7t94nhd99c, 3-oxo-4-cholestene, 3-keto-4-cholestene, cholestenone delta 4
CHEBI:16175
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C
Safety and Handling

Safety and Handling

EINECSNumber : 210-005-1

RTECSNumber : FZ7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only