Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Chlorovalerophenone, 98%

$72.80 - $942.04

Chemical Identifiers

Chemical Name or Material 4'-Chlorovalerophenone
CAS 25017-08-7
Molecular Formula C11H13ClO
Formula Weight 196.68
MDL Number MFCD00018716
Synonym 4-Chlorophenyl-1-pentanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2561109

 
alfa aesar™
B2561109
10g Each for $72.80

AAB2561118

 
alfa aesar™
B2561118
50g Each for $246.28

AAB2561130

 
alfa aesar™
B2561130
250g Each for $942.04
Description & Specifications

Specifications

Formula Weight 196.68
Chemical Name or Material 4'-Chlorovalerophenone
Assay 98%
Molecular Formula C11H13ClO
MDL Number MFCD00018716
CAS 25017-08-7
Synonym 4-Chlorophenyl-1-pentanone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 29° to 33°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.