missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-chlorotoluene, 98%, ACROS Organics™

Manufacturer:  Acros Organics 150210050

 View more versions of this product

Catalog No. AC150210050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Chlorotoluene
51°C
Undesignated
5mL
97.5
Authentic
1.5190 to 1.5210
98%
C7H7Cl
MFCD00000631
15, 2175
NPDACUSDTOMAMK-UHFFFAOYSA-N
1-chloro-4-methylbenzene
7810
126.59
1.0700g/mL
162.0°C
6.0°C to 8.0°C
106-43-4
100.0
Glass bottle
0.01% max.
98%
CH3C6H4Cl
05, 292
PCT, 1-Chloro-4-methylbenzene
CC1=CC=C(C=C1)Cl
126.583
CHEBI:34401
Liquid

Chemical Identifiers

106-43-4
126.583
NPDACUSDTOMAMK-UHFFFAOYSA-N
7810
C7H7Cl
MFCD00000631
PCT, 1-Chloro-4-methylbenzene
CHEBI:34401
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents