missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorothiobenzamide, 97%

Manufacturer:  Alfa Aesar™ A1202109

 View more versions of this product

Catalog No. AAA1202109



Description

Description

Applications
4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

4-Chlorothiobenzamide
10g
C7H6ClNS
606603
OKPUICCJRDBRJT-UHFFFAOYSA-N
4-chlorobenzenecarbothioamide
2734826
97%
123°C to 129°C
2521-24-6
MFCD00040956
4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione
C1=CC(=CC=C1C(=S)N)Cl
171.642
171.65

Chemical Identifiers

2521-24-6
171.642
OKPUICCJRDBRJT-UHFFFAOYSA-N
2734826
C1=CC(=CC=C1C(=S)N)Cl
C7H6ClNS
MFCD00040956
4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione
4-chlorobenzenecarbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents