missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorothiobenzamide, 97%

Manufacturer:  Alfa Aesar™ A1202114

 View more versions of this product

Catalog No. AAA1202114



Description

Description

Applications
4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

4-Chlorothiobenzamide
97%
MFCD00040956
OKPUICCJRDBRJT-UHFFFAOYSA-N
4-chlorobenzenecarbothioamide
2734826
2521-24-6
C7H6ClNS
606603
C1=CC(=CC=C1C(=S)N)Cl
171.642
171.65

Chemical Identifiers

2521-24-6
171.642
OKPUICCJRDBRJT-UHFFFAOYSA-N
4-chlorobenzenecarbothioamide
C7H6ClNS
MFCD00040956
2734826
C1=CC(=CC=C1C(=S)N)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents