Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorothieno[3,2-c]pyridine, 98%

$190.86 - $580.31

Chemical Identifiers

Chemical Name or Material 4-Chlorothieno[3,2-c]pyridine
CAS 27685-94-5
Molecular Formula C7H4ClNS
Formula Weight 169.63
MDL Number MFCD01861830
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH29157MC

 
alfa aesar™
H29157MC
100mg Each for $190.86

AAH29157ME

 
alfa aesar™
H29157ME
500mg Each for $580.31
Description & Specifications

Specifications

Formula Weight 169.63
Chemical Name or Material 4-Chlorothieno[3,2-c]pyridine
Assay 98%
Molecular Formula C7H4ClNS
MDL Number MFCD01861830
CAS 27685-94-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 99° to 101°C
Quantity 100mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.