Please login to your online account to display your discounted pricing

4-(Chlorosulfonyl)benzoic acid, 96%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC405061000

 View more versions of this product

Catalog No. AC405061000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 4-(Chlorosulfonyl)benzoic acid
Color Off-white to beige
Quantity 100g
CAS 10130-89-9
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H5ClO4S
Linear Formula CO2HC6H4SO2Cl
Formula Weight 220.63

Specifications

Chemical Name or Material 4-(Chlorosulfonyl)benzoic acid
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H5ClO4S
Linear Formula CO2HC6H4SO2Cl
Formula Weight 220.63