Please login to your online account to display your discounted pricing

4-Chlorostyrene, Stabilized 99%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  110090500

 View more versions of this product

Catalog No. AC110090500

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 138.6
Physical Form Liquid
Assay ≥98.5%
Quantity 50g
Color Clear colorless to slightly yellow
TSCA TSCA
EINECS Number 214-028-8
Beilstein 05, II, 367
CAS Max % 100.0
Refractive Index 1.5645 to 1.5665 (20°C, 589nm)
Stabilizer 0.1% tert.-butylcatechol
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Name Note 99%
CAS 1073-67-2
Molecular Formula C8H7Cl
Assay Percent Range 99%
Chemical Name or Material 4-Chlorostyrene
MDL Number MFCD00000632
Linear Formula H2CCHC6H4Cl
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Chlorostyrene
Name Note 99%
CAS 1073-67-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7Cl
Linear Formula H2CCHC6H4Cl
Formula Weight 138.6
MDL Number MFCD00000632
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents