Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorosalicylic acid, 98%

$47.26 - $143.61

Chemical Identifiers

Chemical Name or Material 4-Chlorosalicylic acid
CAS 5106-98-9
Molecular Formula C7H5ClO3
Formula Weight 172.57
MDL Number MFCD00002449
Beilstein 2209125
Synonym 4-Chloro-2-hydroxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2252014

 
alfa aesar™
B2252014
25g Each for $47.26

AAB2252022

 
alfa aesar™
B2252022
100g Each for $143.61
Description & Specifications

Specifications

Formula Weight 172.57
Chemical Name or Material 4-Chlorosalicylic acid
Assay 98%
Molecular Formula C7H5ClO3
MDL Number MFCD00002449
CAS 5106-98-9
Beilstein 2209125
Synonym 4-Chloro-2-hydroxybenzoic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈215°C (dec.)
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.