missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloroquinoline-6-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H54085MD

 View more versions of this product

Catalog No. AAH54085MD



Description

Description

Applications
4-Chloroquinoline-6-boronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-Chloroquinoline-6-boronic acid pinacol ester
1201844-73-6
MFCD16987803
ANPKKHFSNJKOAM-UHFFFAOYSA-N
4-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
53363436
96%
250mg
C15H17BClNO2
4-chloro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=CN=C3C=C2)Cl
289.566
289.56

Chemical Identifiers

1201844-73-6
289.566
ANPKKHFSNJKOAM-UHFFFAOYSA-N
53363436
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=CN=C3C=C2)Cl
C15H17BClNO2
MFCD16987803
4-chloro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline
4-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents