Please login to your online account to display your discounted pricing

4-Chloroquinaldine, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Chloroquinaldine
Assay Percent Range 99%
Molecular Formula C10H8ClN
Formula Weight 177.63
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC165550050

 
thermo fisher scientific
AC165550050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC165550250

  SDS
thermo fisher scientific
AC165550250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.63
Quantity 5g
Chemical Name or Material 4-Chloroquinaldine
Assay Percent Range 99%
CAS 4295-06-1
Molecular Formula C10H8ClN