Please login to your online account to display your discounted pricing

4-Chloroquinaldine, 99%, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  AC165550250

 View more versions of this product

Catalog No. AC165550250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 4-Chloroquinaldine
Quantity 25g
CAS 4295-06-1
Assay Percent Range 99%
Molecular Formula C10H8ClN
Formula Weight 177.63

Specifications

Chemical Name or Material 4-Chloroquinaldine
Assay Percent Range 99%
Molecular Formula C10H8ClN
Formula Weight 177.63