missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloropyridine hydrochloride, 98+%

Manufacturer:  Alfa Aesar™ A1562822

 View more versions of this product

Catalog No. AAA1562822



Description

Description

Applications
It is employed as a catalyst in the synthesis of fluorazone derivatives by one-pot pyrrolation/cyclization of anthranilic acids.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2
198°C (388°F)
~210°C (sublimation)
7379-35-3
MFCD00012829
4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride
C1=CN=CC=C1Cl.Cl
150.002
150.01
4-Chloropyridine hydrochloride
Weak
100g
C5H5Cl2N
3684904
XGAFCCUNHIMIRV-UHFFFAOYSA-N
4-chloropyridine;hydrochloride
81852
≥98%

Chemical Identifiers

7379-35-3
150.002
XGAFCCUNHIMIRV-UHFFFAOYSA-N
81852
C1=CN=CC=C1Cl.Cl
C5H5Cl2N
MFCD00012829
4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride
4-chloropyridine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280f-P305+P351+P338
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : US6300000

TSCA : TSCA

Documents