missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloropyridine hydrochloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 110031000

 View more versions of this product

Catalog No. AC110031000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chloropyridine hydrochloride
7379-35-3
100.0
Beige-Yellow to White
Authentic
Plastic Bottle
(C5H4N)Cl
100 g
4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride
XGAFCCUNHIMIRV-UHFFFAOYSA-N
4-chloropyridine;hydrochloride
81852
97%
97%
96.0
210.0°C
198°C
96% min. (HPLC)
C5H5Cl2N
MFCD00012829
20, 231
Solubility in water: soluble
[H+].[Cl-].ClC1=CC=NC=C1
150.00
150.01
Crystalline Powder

Chemical Identifiers

7379-35-3
150.00
XGAFCCUNHIMIRV-UHFFFAOYSA-N
81852
[H+].[Cl-].ClC1=CC=NC=C1
C5H5Cl2N
MFCD00012829
4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride
4-chloropyridine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 230-946-1

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only