Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorophenylurea, 98%

$53.35 - $664.49

Chemical Identifiers

Chemical Name or Material 4-Chlorophenylurea
CAS 140-38-5
Molecular Formula C7H7ClN2O
Formula Weight 170.6
MDL Number MFCD00014788
Beilstein 908492
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2509006

  SDS
alfa aesar™
B2509006
5g Each for $53.35

AAB2509014

  SDS
alfa aesar™
B2509014
25g Each for $206.43

AAB2509022

  SDS
alfa aesar™
B2509022
100g Each for $664.49
Description & Specifications

Specifications

Formula Weight 170.6
MDL Number MFCD00014788
RTECS Number YS6127000
Quantity 5g
Molecular Formula C7H7ClN2O
CAS 140-38-5
Assay 98%
Beilstein 908492
Melting Point 204° to 205°C
Chemical Name or Material 4-Chlorophenylurea