Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Chlorophenylsulfonyl)acetonitrile, 98+%

Chemical Identifiers

Chemical Name or Material (4-Chlorophenylsulfonyl)acetonitrile
CAS 1851-09-8
Molecular Formula C8H6ClNO2S
Formula Weight 215.66
MDL Number MFCD00045626
UN Number UN3439
Beilstein 1819678
Synonym (4-Chlorobenzenesulfonyl)acetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1451003

  SDS
alfa aesar™
A1451003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1451006

  SDS
alfa aesar™
A1451006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1451014

  SDS
alfa aesar™
A1451014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.66
Chemical Name or Material (4-Chlorophenylsulfonyl)acetonitrile
Assay ≥98%
Molecular Formula C8H6ClNO2S
MDL Number MFCD00045626
CAS 1851-09-8
UN Number UN3439
Beilstein 1819678
Synonym (4-Chlorobenzenesulfonyl)acetonitrile
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Melting Point 169° to 173°C
Quantity 1g

Applications
(4-Chlorophenylsulfonyl)acetonitrile react with trimethoxymethyl-benzene, and gives 2-(4-chloro-benzenesulfonyl)-3-methoxy-3-phenyl-acrylonitrile.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.