missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Chlorophenylsulfonyl)acetonitrile, 98+%

Manufacturer:  Alfa Aesar™ A1451014

 View more versions of this product

Catalog No. AAA1451014



Description

Description

Applications
(4-Chlorophenylsulfonyl)acetonitrile react with trimethoxymethyl-benzene, and gives 2-(4-chloro-benzenesulfonyl)-3-methoxy-3-phenyl-acrylonitrile.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

(4-Chlorophenylsulfonyl)acetonitrile
25g
C8H6ClNO2S
UN3439
4-chlorophenylsulfonylacetonitrile, 2-4-chlorophenyl sulfonyl acetonitrile, 4-chlorophenyl cyanomethyl sulfone, 2-4-chlorobenzenesulfonyl acetonitrile, 4-chlorobenzenesulphonylacetonitrile, 4-chlorobenzenesulfonyl acetonitrile, 4-chlorobenzenesulphonyl acetonitrile, 4-chlorophenyl sulfonyl acetonitrile, 2-4-chlorophenyl sulfonylacetonitrile, 4-chlorophenylsulfonyl acetonitrile
C1=CC(=CC=C1S(=O)(=O)CC#N)Cl
215.651
215.66
169°C to 173°C
1851-09-8
MFCD00045626
1819678
HAQGVGPNKGGSMK-UHFFFAOYSA-N
2-(4-chlorophenyl)sulfonylacetonitrile
735829
≥98%

Chemical Identifiers

1851-09-8
215.651
HAQGVGPNKGGSMK-UHFFFAOYSA-N
735829
C1=CC(=CC=C1S(=O)(=O)CC#N)Cl
C8H6ClNO2S
MFCD00045626
4-chlorophenylsulfonylacetonitrile, 2-4-chlorophenyl sulfonyl acetonitrile, 4-chlorophenyl cyanomethyl sulfone, 2-4-chlorobenzenesulfonyl acetonitrile, 4-chlorobenzenesulphonylacetonitrile, 4-chlorobenzenesulfonyl acetonitrile, 4-chlorobenzenesulphonyl acetonitrile, 4-chlorophenyl sulfonyl acetonitrile, 2-4-chlorophenyl sulfonylacetonitrile, 4-chlorophenylsulfonyl acetonitrile
2-(4-chlorophenyl)sulfonylacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents