Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Chlorophenylsulfonyl)acetone, 98%

$24.42 - $376.69

Chemical Identifiers

Chemical Name or Material (4-Chlorophenylsulfonyl)acetone
CAS 5000-48-6
Molecular Formula C9H9ClO3S
Formula Weight 232.69
MDL Number MFCD00018663
Beilstein 2051464
Synonym (4-Chlorobenzenesulfonyl)acetone; 1-(4-Chlorophenylsulfonyl)-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1485803

  SDS
alfa aesar™
A1485803
1g Each for $24.42

AAA1485806

  SDS
alfa aesar™
A1485806
5g Each for $88.59

AAA1485814

  SDS
alfa aesar™
A1485814
25g Each for $376.69
Description & Specifications

Specifications

Formula Weight 232.69
Melting Point 78° to 82°C
Chemical Name or Material (4-Chlorophenylsulfonyl)acetone
Quantity 1g
Assay 98%
Molecular Formula C9H9ClO3S
MDL Number MFCD00018663
CAS 5000-48-6
Beilstein 2051464
Synonym (4-Chlorobenzenesulfonyl)acetone; 1-(4-Chlorophenylsulfonyl)-2-propanone