Alfa Aesar™ (4-Chlorophenylsulfonyl)acetone, 98%
Manufacturer:  Alfa Aesar™ A1485803
 View more versions of this product
Catalog No. AAA1485803
Specifications
Specifications
(4-Chlorophenylsulfonyl)acetone
1g
C9H9ClO3S
2051464
BRDBHPZILGTBFY-UHFFFAOYSA-N
1-(4-chlorophenyl)sulfonylpropan-2-one
521269
98%
78°C to 82°C
5000-48-6
MFCD00018663
4-chlorophenylsulfonylacetone, 1-4-chlorophenyl sulfonyl acetone, 1-4-chlorophenyl sulfonyl propan-2-one, 4-chlorobenzenesulphonylacetone, p-chlorophenylsulfonylacetone, 1-4-chlorobenzenesulfonyl propan-2-one, 4-chlorophenylsulfonyl acetone, 1-4-chlorophenyl sulfonylpropan-2-one, 1-4-chlorophenyl sulfonyl propan-2-one, maybridge1_004685
CC(=O)CS(=O)(=O)C1=CC=C(C=C1)Cl
232.678
232.69
Chemical Identifiers
5000-48-6
232.678
BRDBHPZILGTBFY-UHFFFAOYSA-N
521269
CC(=O)CS(=O)(=O)C1=CC=C(C=C1)Cl
C9H9ClO3S
MFCD00018663
4-chlorophenylsulfonylacetone, 1-4-chlorophenyl sulfonyl acetone, 1-4-chlorophenyl sulfonyl propan-2-one, 4-chlorobenzenesulphonylacetone, p-chlorophenylsulfonylacetone, 1-4-chlorobenzenesulfonyl propan-2-one, 4-chlorophenylsulfonyl acetone, 1-4-chlorophenyl sulfonylpropan-2-one, 1-4-chlorophenyl sulfonyl propan-2-one, maybridge1_004685
1-(4-chlorophenyl)sulfonylpropan-2-one
SDS Documents