Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Chlorophenylsulfonyl)acetic acid, 98%

$153.69 - $462.58

Chemical Identifiers

Chemical Name or Material (4-Chlorophenylsulfonyl)acetic acid
CAS 3405-89-8
Molecular Formula C8H7ClO4S
Formula Weight 234.66
MDL Number MFCD00201948
Synonym (4-Chlorobenzenesulfonyl)acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27414MC

  SDS
alfa aesar™
H27414MC
100mg Each for $153.69

AAH27414ME

  SDS
alfa aesar™
H27414ME
500mg Each for $462.58
Description & Specifications

Specifications

Formula Weight 234.66
MDL Number MFCD00201948
Assay 98%
Health Hazard 1 Warning
Chemical Name or Material (4-Chlorophenylsulfonyl)acetic acid
Quantity 100mg
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 3405-89-8
Melting Point ≈140°C
Synonym (4-Chlorobenzenesulfonyl)acetic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H7ClO4S
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.