Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Chlorophenylsulfonyl)acetamidoxime

$327.59 - $327.59

Chemical Identifiers

Chemical Name or Material (4-Chlorophenylsulfonyl)acetamidoxime
CAS 70661-64-2
Molecular Formula C8H9ClN2O3S
Formula Weight 248.69g/mol
MDL Number MFCD05864482
Synonym 2-[(4-Chlorophenyl)sulfonyl]-N-hydroxyethanimidamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5068106

 
alfa aesar™
H50681-06
5g Each for $327.59
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material (4-Chlorophenylsulfonyl)acetamidoxime
CAS 70661-64-2
Molecular Formula C8H9ClN2O3S
Formula Weight 248.69g/mol
MDL Number MFCD05864482
Synonym 2-[(4-Chlorophenyl)sulfonyl]-N-hydroxyethanimidamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.