Please login to your online account to display your discounted pricing

4-Chlorophenylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC377408000

 View more versions of this product

Catalog No. AC377408000

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Store in a dry place. Store in a closed container. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Health Hazard 2 GHS H Statement
Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
Physical Form Solution
Formula Weight 215.76
Quantity 800mL
Color Light gray to yellow to brown-black
Packaging AcroSeal™ Glass Bottle
Concentration 0.9 to 1.2M
Density 0.8500g/mL
Linear Formula Cl(C6H4)MgBr
Storage Information Magnesium salts may crystallize below 20°C
Health Hazard 1 Danger
Chemical Name or Material 4-Chlorophenylmagnesium bromide
MDL Number MFCD00009926
CAS 873-77-8
CAS 60-29-7
CAS Min % 22.85
CAS Min % 69.54
CAS Max % 77.15
CAS Max % 30.46
Name Note 1M Solution in Diethyl Ether
Molecular Formula C6H4BrClMg

Specifications

Chemical Name or Material 4-Chlorophenylmagnesium bromide
Name Note 1M Solution in Diethyl Ether
CAS 873-77-8
CAS Min % 22.85
CAS Max % 30.46
CAS 60-29-7
CAS Min % 69.54
CAS Max % 77.15
Molecular Formula C6H4BrClMg
Linear Formula Cl(C6H4)MgBr
Formula Weight 215.76
MDL Number MFCD00009926
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Store in a dry place. Store in a closed container. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS H Statement
Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.

SDS & Safety Documents