Please login to your online account to display your discounted pricing

4-Chlorophenylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC377401000

 View more versions of this product

Catalog No. AC377401000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Store in a dry place. Store in a closed container. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Formula Weight 215.76
Physical Form Solution
Quantity 100mL
Color Light gray to yellow to brown-black
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.8500g/mL
Concentration 0.9 to 1.2M
Health Hazard 1 Danger
Chemical Name or Material 4-Chlorophenylmagnesium bromide
CAS Max % 77.15
CAS Max % 30.46
CAS Min % 69.54
CAS 60-29-7
CAS Min % 22.85
Storage Information Magnesium salts may crystallize below 20°C
Linear Formula Cl(C6H4)MgBr
CAS 873-77-8
Name Note 1M Solution in Diethyl Ether
Molecular Formula C6H4BrClMg
MDL Number MFCD00009926

Specifications

Chemical Name or Material 4-Chlorophenylmagnesium bromide
Name Note 1M Solution in Diethyl Ether
CAS 873-77-8
CAS Min % 22.85
CAS Max % 30.46
CAS 60-29-7
CAS Min % 69.54
CAS Max % 77.15
Molecular Formula C6H4BrClMg
Linear Formula Cl(C6H4)MgBr
Formula Weight 215.76
MDL Number MFCD00009926
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Store in a dry place. Store in a closed container. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS H Statement
Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.

SDS & Safety Documents